Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner, Oil/Grease Remover, 1 Gallon, 4/CT, White
SDP
Catalog No. 5000790518
Encompass_Preferred
Floor cleaner/oil and grease remover is a nonionic oil and grease remover for all resilient and synthetic flooring. Highly effective, solvent-free formula emulsifies dirt quickly. Neutral-pH and butyl-free formula is gentle enough to use on resilient or rubber flooring. Cleaner is an integral part of a comprehensive floor-care program for cleaning, stripping and sealing rubber floors. Floor cleaner is perfect for use as floor cleaner and grease remover for all resilient and synthetic rubber studded flooring, and can also be used on stone, marble and other hard surfaces.
Catalog No. 50-007-90518 Supplier Enterprise Technology Solutions Supplier No. DVO94512759
Floor cleaner/oil and grease remover is a nonionic oil and grease remover for all resilient and synthetic flooring. Highly effective, solvent-free formula emulsifies dirt quickly. Neutral-pH and butyl-free formula is gentle enough to use on resilient or rubber flooring. Cleaner is an integral part of a comprehensive floor-care program for cleaning, stripping and sealing rubber floors. Floor cleaner is perfect for use as floor cleaner and grease remover for all resilient and synthetic rubber studded flooring, and can also be used on stone, marble and other hard surfaces.