Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner, Revive, 5 Gallon Envirobox, Pink
SDP
Catalog No. 5000790529
Encompass_Preferred
Revive Cleaner/Maintainer features an easy-to-use design to clean and maintain floors in a single step and does not require burnishing after each application. Fast-acting formula dries immediately when used in an automatic scrubber to save labor and time. Cleaner with a slightly sweet scent can be used through an auto scrubber or with mop and bucket. Dilution ratios are 1:32 and 1:64.
Catalog No. 50-007-90529 Supplier Enterprise Technology Solutions Supplier No. DVO101109868
Revive Cleaner/Maintainer features an easy-to-use design to clean and maintain floors in a single step and does not require burnishing after each application. Fast-acting formula dries immediately when used in an automatic scrubber to save labor and time. Cleaner with a slightly sweet scent can be used through an auto scrubber or with mop and bucket. Dilution ratios are 1:32 and 1:64.