Learn More
Enterprise Technology Solutions Diversey Care Floor Finish,High Mileage,RTU,Envirobox,5 Gallon,White
SDP
Catalog No. 5000790164
Encompass_Preferred
High-solids, low-odor floor finish features an advanced formula to deliver excellent gloss build and outstanding durability, even in the heaviest traffic. Non-yellowing design maintains excellent clear color. Extended recoat and stripping cycles yield labor savings and overall improved productivity. Fast-drying design allows you to install up to five coats in one night. Specially designed, higher solids formula builds durability over time. Product does not contain any intentionally added PFAS (Perfluoroalkyl and Polyfluoroalkyl substances).
Catalog No. 50-007-90164 Supplier Enterprise Technology Solutions Supplier No. DVO101110008
High-solids, low-odor floor finish features an advanced formula to deliver excellent gloss build and outstanding durability, even in the heaviest traffic. Non-yellowing design maintains excellent clear color. Extended recoat and stripping cycles yield labor savings and overall improved productivity. Fast-drying design allows you to install up to five coats in one night. Specially designed, higher solids formula builds durability over time. Product does not contain any intentionally added PFAS (Perfluoroalkyl and Polyfluoroalkyl substances).