Learn More
Enterprise Technology Solutions Diversey Care Floor Finish, Matte, Carefree, 5 Gallon, White
SDP
Supplier:  Enterprise Technology Solutions DVO5104757
Encompass_Preferred
Carefree Matte Floor Finish features a versatile, high-performance design to work with a wide variety of substrates and in many different maintenance methods. Low-gloss characteristics help hide blemishes, cut lines, traffic lanes and dirt embedment to provide an even, clean look over a longer period of time. Floor finish meets ASTM D-2047 slip-resistance requirements.
Catalog No. 50-007-90076