Learn More
Enterprise Technology Solutions Diversey Care Floor Finish,Vectra,RTU,Ammonia,5 Gal Envirobox,Off-White
SDP
Catalog No. 5000790478
Encompass_Preferred
Vectra Floor Finish provides diamond-like brilliance, clarity and durability. Advantages include a faster, early cure time and better buff response and improved dirt-resistance over other scrub and recoat products. Floor finish with an ammonia scent resists dirt embedment, black heel marks, scratching and scuffing. It responds well to 175 through 1500 RPM equipment and will work with UHS equipment.
Catalog No. 50-007-90478 Supplier Enterprise Technology Solutions Supplier No. DVO101109988
Vectra Floor Finish provides diamond-like brilliance, clarity and durability. Advantages include a faster, early cure time and better buff response and improved dirt-resistance over other scrub and recoat products. Floor finish with an ammonia scent resists dirt embedment, black heel marks, scratching and scuffing. It responds well to 175 through 1500 RPM equipment and will work with UHS equipment.