Learn More
Enterprise Technology Solutions Diversey Care Floor Pad,180 Grit,5",12/PK,4PK/CT,OE
SDP
Catalog No. 5000790297
Encompass_Preferred
Bona Diamond Abrasive Discs deliver consistent scratch patterns across the entire floor, creating the best surface for optimal adhesion. These revolutionary discs prepare all polyurethane-finished floors for recoating, including hard-to-abrade ALOX and ceramic prefinished surfaces. Engineered to last 12 times longer than screens and other inter-coat abrasives, they provide exceptional durability and value. Their unique construction allows them to conform to the floor, ensuring complete abrasion and smoothing for a flawless, scratch-free finish on raw wood or recoats. Each set covers 4,000 to 6,000 square feet during inter-coat abrasion, reducing downtime and replacement needs. With no visible swirl marks, a consistent scratch pattern and improved adhesion for recoating, Bona Diamond Abrasives set a new standard in surface preparation.
Catalog No. 50-007-90297 Supplier Enterprise Technology Solutions Supplier No. DVOAASDIAMOND18
Bona Diamond Abrasive Discs deliver consistent scratch patterns across the entire floor, creating the best surface for optimal adhesion. These revolutionary discs prepare all polyurethane-finished floors for recoating, including hard-to-abrade ALOX and ceramic prefinished surfaces. Engineered to last 12 times longer than screens and other inter-coat abrasives, they provide exceptional durability and value. Their unique construction allows them to conform to the floor, ensuring complete abrasion and smoothing for a flawless, scratch-free finish on raw wood or recoats. Each set covers 4,000 to 6,000 square feet during inter-coat abrasion, reducing downtime and replacement needs. With no visible swirl marks, a consistent scratch pattern and improved adhesion for recoating, Bona Diamond Abrasives set a new standard in surface preparation.