Learn More
Enterprise Technology Solutions Diversey Care Floor Stripper,Hvy-dty,Cherry Almond,Envirobox,5 Gal,Straw
SDP
Catalog No. 5000790904
Encompass_Preferred
Pro Strip Heavy Duty Floor Stripper is a professional-strength stripper designed for the removal of "tough to strip" finishes and sealers. That means it is formulated for effective stripping the first time, reducing the need for additional restrips. Floor stripper delivers fast penetration of built-up finishes and sealer layers and is specially formulated to penetrate and emulsify the toughest finishes and sealers. Variable dilution rates (1:4 and 1:10) using cool water offers a cost-effective solution. Pleasant cherry almond fragrance reduces complaints regarding floor-stripping odors.
Catalog No. 50-007-90904 Supplier Enterprise Technology Solutions Supplier No. DVO95032360
Pro Strip Heavy Duty Floor Stripper is a professional-strength stripper designed for the removal of "tough to strip" finishes and sealers. That means it is formulated for effective stripping the first time, reducing the need for additional restrips. Floor stripper delivers fast penetration of built-up finishes and sealer layers and is specially formulated to penetrate and emulsify the toughest finishes and sealers. Variable dilution rates (1:4 and 1:10) using cool water offers a cost-effective solution. Pleasant cherry almond fragrance reduces complaints regarding floor-stripping odors.