Learn More
Enterprise Technology Solutions Diversey Care Floor Stripper,RTD,Low-odor,Concentrated,5L,Amber
SDP
Catalog No. 5000790426
Encompass_Preferred
Part of the Diversey RTD System, this Pro Strip SC High Efficiency Floor Stripper quickly bites through layers of floor finish on resilient flooring. It decreases downtime and labor costs while reducing the odors associated with floor stripping. This highly concentrated, low-odor floor stripper is not formulated with ammonia or butyl. Dilution rations are 1:13 and 1:20. Powerful floor stripper with a solvent scent meets Green Seal GS-40 standard for industrial and institutional floor-care products.
Catalog No. 50-007-90426 Supplier Enterprise Technology Solutions Supplier No. DVO95547312
Part of the Diversey RTD System, this Pro Strip SC High Efficiency Floor Stripper quickly bites through layers of floor finish on resilient flooring. It decreases downtime and labor costs while reducing the odors associated with floor stripping. This highly concentrated, low-odor floor stripper is not formulated with ammonia or butyl. Dilution rations are 1:13 and 1:20. Powerful floor stripper with a solvent scent meets Green Seal GS-40 standard for industrial and institutional floor-care products.