Learn More
Enterprise Technology Solutions Diversey Care Floor Stripper, Ultra, Heavy-Duty, 5 Gallon, Red
SDP
Catalog No. 5000789885
Encompass_Preferred
Heavy-duty floor stripper features a powerful formula to quickly remove tough-to-strip finishes and sealers with fewer passes. Advanced design effectively strips the first time to reduce the need for additional re-strips. Fast-acting solution quickly penetrates built-up finishes and sealer layers and is specially formulated to penetrate and emulsify the toughest UHS burnished finishes. Cost-effective stripper offers variable dilution rates, utilizing cool water. Pleasant cherry almond fragrance reduces complaints regarding floor-stripping odors. Floor stripper is designed for floor finish removal on VCT, terrazzo and concrete flooring.
Catalog No. 50-007-89885 Supplier Enterprise Technology Solutions Supplier No. DVO95386176
Heavy-duty floor stripper features a powerful formula to quickly remove tough-to-strip finishes and sealers with fewer passes. Advanced design effectively strips the first time to reduce the need for additional re-strips. Fast-acting solution quickly penetrates built-up finishes and sealer layers and is specially formulated to penetrate and emulsify the toughest UHS burnished finishes. Cost-effective stripper offers variable dilution rates, utilizing cool water. Pleasant cherry almond fragrance reduces complaints regarding floor-stripping odors. Floor stripper is designed for floor finish removal on VCT, terrazzo and concrete flooring.