Learn More
Enterprise Technology Solutions Diversey Care FloorPads,f/Unprotected,Clean&Polish,Dmnds,Round,20",2/PK,GN
SDP
Catalog No. 5000790906
Encompass_Preferred
Twister Diamond Pads offer an innovative method for daily floor maintenance. Easily clean and polish a variety of unprotected floors with floor pads prepared with microscopic diamonds. The diamond-impregnated side comes with color coding for easier identification and works as a wear indicator. Enjoy cleaner and glossier floors with every use.
Catalog No. 50-007-90906 Supplier Enterprise Technology Solutions Supplier No. DVODD5871037
Twister Diamond Pads offer an innovative method for daily floor maintenance. Easily clean and polish a variety of unprotected floors with floor pads prepared with microscopic diamonds. The diamond-impregnated side comes with color coding for easier identification and works as a wear indicator. Enjoy cleaner and glossier floors with every use.