Learn More
Enterprise Technology Solutions Diversey Care Glass Cleaner, Ammoniated, Glance, 1.5 Liter, 2/CT, Blue
SDP
Catalog No. 5000790288
Encompass_Preferred
Glass/multi-surface cleaner features a quick-drying, ammoniated formula to leave a streak-free shine on surfaces. Ready-to-use formula makes cleaning quick and simple. Concentrated cleaner provides cost-efficient cleaning performance. Versatile design also cuts through grease, grime, soap film, finger marks and smoke film. Glass/multi-surface cleaner is perfect for glass, mirrors, windows, sinks, countertops, stainless steel and more.
Catalog No. 50-007-90288 Supplier Enterprise Technology Solutions Supplier No. DVO93063402
Glass/multi-surface cleaner features a quick-drying, ammoniated formula to leave a streak-free shine on surfaces. Ready-to-use formula makes cleaning quick and simple. Concentrated cleaner provides cost-efficient cleaning performance. Versatile design also cuts through grease, grime, soap film, finger marks and smoke film. Glass/multi-surface cleaner is perfect for glass, mirrors, windows, sinks, countertops, stainless steel and more.