Learn More
Enterprise Technology Solutions Diversey Care Glass Cleaner,Kosher,1/2oz,100 Pouches/CT,White
SDP
Catalog No. 5000790424
Encompass_Preferred
For sparkling clean glassware and better-tasting beer, Beer Clean Glass Cleaner provides thoroughly cleaned glassware with no odor, aftertaste or residue. Powdered compound cleans in hot-cold, hard or soft water. It is simple to use. Concentrated formula means quick cleaning. Odorless, Kosher detergent system provides excellent rinsing results. Use with manual brush cleaning.
Catalog No. 50-007-90424 Supplier Enterprise Technology Solutions Supplier No. DVO990221
For sparkling clean glassware and better-tasting beer, Beer Clean Glass Cleaner provides thoroughly cleaned glassware with no odor, aftertaste or residue. Powdered compound cleans in hot-cold, hard or soft water. It is simple to use. Concentrated formula means quick cleaning. Odorless, Kosher detergent system provides excellent rinsing results. Use with manual brush cleaning.