Learn More
Enterprise Technology Solutions Diversey Care Hand Sanitizer, f/IntelliCare, RTU, 1.3L, Clear
SDP
Catalog No. 5000790422
Encompass_Preferred
Made for Diversey IntelliCare Dispensers, this Soft Care Instant Hand Sanitizer AF offers an alcohol-free formulation for use where alcohol-based hand sanitizers are a concern. Benzalkonium chloride-based hand sanitizer is enriched with skin-conditioning emollients to keep skin soft and protected against irritation but leaves no sticky residues. Safe, quick and effective hand rub helps reduce the spread of germs. FDA and NSF E-3 certified for use in food-handling establishments, it is great for use in restrooms and public areas found in retail, food-handling establishments, schools, prisons, healthcare facilities, offices and fitness centers. Dye-free hand sanitizer reduces potential skin irritation and allergic reactions for users with sensitive skin.
Catalog No. 50-007-90422 Supplier Enterprise Technology Solutions Supplier No. DVO100961733
Made for Diversey IntelliCare Dispensers, this Soft Care Instant Hand Sanitizer AF offers an alcohol-free formulation for use where alcohol-based hand sanitizers are a concern. Benzalkonium chloride-based hand sanitizer is enriched with skin-conditioning emollients to keep skin soft and protected against irritation but leaves no sticky residues. Safe, quick and effective hand rub helps reduce the spread of germs. FDA and NSF E-3 certified for use in food-handling establishments, it is great for use in restrooms and public areas found in retail, food-handling establishments, schools, prisons, healthcare facilities, offices and fitness centers. Dye-free hand sanitizer reduces potential skin irritation and allergic reactions for users with sensitive skin.