Learn More
Enterprise Technology Solutions Diversey Care Hand Sanitizer Refill,Foam,f/IntelliCare Disp,1.2L,6/CT,CL
SDP
Catalog No. 5000790566
Encompass_Preferred
Soft Care Impact Foam Instant Hand Sanitizer contains skin-softening emollients that leave hands soft and with no sticky residue. They protect against irritation for even the most frequent users. Fragrance-free and dye-free, this hand sanitizer meets CDC, WHO, APIC and PHAC hand hygiene recommendations for healthcare personnel hand rubs. It is EcoLogo Certified, NSF E-3 certified for use in food handling establishments and a USDA Certified Biobased product. Made with a 76 percent alcohol-based formula, commercial hand sanitizer is great for use in restrooms, food-handling facilities, healthcare settings, offices and fitness centers. The 1200 ml. refill is designed for use in both the Diversey IntelliCare Hybrid Dispenser and IntelliCare Manual Dispenser II.
Catalog No. 50-007-90566 Supplier Enterprise Technology Solutions Supplier No. DVO100907873
Soft Care Impact Foam Instant Hand Sanitizer contains skin-softening emollients that leave hands soft and with no sticky residue. They protect against irritation for even the most frequent users. Fragrance-free and dye-free, this hand sanitizer meets CDC, WHO, APIC and PHAC hand hygiene recommendations for healthcare personnel hand rubs. It is EcoLogo Certified, NSF E-3 certified for use in food handling establishments and a USDA Certified Biobased product. Made with a 76 percent alcohol-based formula, commercial hand sanitizer is great for use in restrooms, food-handling facilities, healthcare settings, offices and fitness centers. The 1200 ml. refill is designed for use in both the Diversey IntelliCare Hybrid Dispenser and IntelliCare Manual Dispenser II.