Learn More
Enterprise Technology Solutions Diversey Care LinoSAFE Linoleum/Stone Stripper, 5 Gallon Envirobox, Clear
SDP
Catalog No. 5000830988
Encompass_Preferred
LinoSAFE Linoleum/Stone Stripper efficiently removes existing floor finish on linoleum, rubber, marble and other sensitive substrates. Concentrate will not damage or discolor sensitive substrates with proper use. Apply with a mop. Dilution ratio is 1:4.
Catalog No. 50-008-30988 Supplier Enterprise Technology Solutions Supplier No. DVO101104658
LinoSAFE Linoleum/Stone Stripper efficiently removes existing floor finish on linoleum, rubber, marble and other sensitive substrates. Concentrate will not damage or discolor sensitive substrates with proper use. Apply with a mop. Dilution ratio is 1:4.