Learn More
Enterprise Technology Solutions Diversey Care Maintainer/Rejuvenator, Revive Plus, 32 oz, 6/CT, Red
SDP
Catalog No. 5000830921
Encompass_Preferred
Revive Plus SC Maintainer/Rejuvenator features a unique formula that repairs microscopic scratches to maintain a glossy look for your floors. Gentle ingredients allow safe use through daily cleanings without browning out. Advanced solution enhances your floor's appearance when burnished with 1500 to 2000 plus rpm propane, battery or electric burnishers. Cleaner/maintainer is designed for use as a hard-surface maintainer.
Catalog No. 50-008-30921 Supplier Enterprise Technology Solutions Supplier No. DVO92974600
Revive Plus SC Maintainer/Rejuvenator features a unique formula that repairs microscopic scratches to maintain a glossy look for your floors. Gentle ingredients allow safe use through daily cleanings without browning out. Advanced solution enhances your floor's appearance when burnished with 1500 to 2000 plus rpm propane, battery or electric burnishers. Cleaner/maintainer is designed for use as a hard-surface maintainer.