Learn More
Enterprise Technology Solutions Diversey Care Mop Pads,Single-Use,Wet/Dry,11.4"x19.3"x20.5",50/PK,10/CT,WE
SDP
Catalog No. 5000831283
Encompass_Preferred
EasyMop Single-Use Mop Pads deliver hygienic cleaning that only single-use mops can offer. Single-use material provides superior cleaning performance , wet or dry, and removes germs to help prevent contamination in critical areas. Disposable mop pads can hold and distribute more liquid than a typical disposable mop, covering approximately 180 square feet. Sheets do not bind with quats, or other chemistries, for effective disinfection. For best results, use with TASKI Jon Master UltraPlus T-Handle and UltraPlus mop frames (not included).
Catalog No. 50-008-31283 Supplier Enterprise Technology Solutions Supplier No. DVOD1232213
Add to Cart
Add to Cart
EasyMop Single-Use Mop Pads deliver hygienic cleaning that only single-use mops can offer. Single-use material provides superior cleaning performance , wet or dry, and removes germs to help prevent contamination in critical areas. Disposable mop pads can hold and distribute more liquid than a typical disposable mop, covering approximately 180 square feet. Sheets do not bind with quats, or other chemistries, for effective disinfection. For best results, use with TASKI Jon Master UltraPlus T-Handle and UltraPlus mop frames (not included).