Learn More
Enterprise Technology Solutions Diversey Care Neutral Cleaner, Smart Dose, Citrus Scent, 1.4L, Orange
SDP
Catalog No. 5000831310
Encompass_Preferred
Concentrated neutral cleaner is formulated for everyday cleaning of floors and other hard surfaces. Use a daily floor cleaner and general cleaner. This product does not dull floor finishes and does not require rinsing to deliver labor savings. Nonalkaline formulation dries film-free. Low-foaming formula allows less emptying of recovery tanks. Use as spray-and-wipe application, in auto scrubber or in mop and bucket. Concentrated formulations deliver excellent cleaning performance for just pennies per diluted gallon. Bottle and pump are designed not to waste concentrate because they evacuate 99.9 percent of the liquid. Automatic measurements deliver precise doses per application, providing consistent cleaning. Easy-to-use cleaner is orange in color and has a citrus scent. This neutral cleaner is part of the Diversey SmartDose system, effectively delivering accurate doses of concentrated product simply, safely and swiftly.
Catalog No. 50-008-31310 Supplier Enterprise Technology Solutions Supplier No. DVO95122613
Concentrated neutral cleaner is formulated for everyday cleaning of floors and other hard surfaces. Use a daily floor cleaner and general cleaner. This product does not dull floor finishes and does not require rinsing to deliver labor savings. Nonalkaline formulation dries film-free. Low-foaming formula allows less emptying of recovery tanks. Use as spray-and-wipe application, in auto scrubber or in mop and bucket. Concentrated formulations deliver excellent cleaning performance for just pennies per diluted gallon. Bottle and pump are designed not to waste concentrate because they evacuate 99.9 percent of the liquid. Automatic measurements deliver precise doses per application, providing consistent cleaning. Easy-to-use cleaner is orange in color and has a citrus scent. This neutral cleaner is part of the Diversey SmartDose system, effectively delivering accurate doses of concentrated product simply, safely and swiftly.