Learn More
Enterprise Technology Solutions Diversey Care Odor Eliminator,Fresh Scent,1 Gallon,4/CT,Tan
SDP
Catalog No. 5000831500
Encompass_Preferred
Good Sense Odor Eliminator is a residual-action odor counteractant that utilizes nonpathogenic bacteria and the enzymes they produce to help remove foul odors caused by urine, feces, blood, vomit, food spills, garbage and grease. It is perfect for odor control and residual odor digesting while leaving behind a fresh scent. Good Sense helps prevent drain clogs while removing odors and is safe to use on all washable surfaces. Formula is EcoLogo certified and kosher.
Catalog No. 50-008-31500 Supplier Enterprise Technology Solutions Supplier No. DVO94496154
Good Sense Odor Eliminator is a residual-action odor counteractant that utilizes nonpathogenic bacteria and the enzymes they produce to help remove foul odors caused by urine, feces, blood, vomit, food spills, garbage and grease. It is perfect for odor control and residual odor digesting while leaving behind a fresh scent. Good Sense helps prevent drain clogs while removing odors and is safe to use on all washable surfaces. Formula is EcoLogo certified and kosher.