Learn More
Enterprise Technology Solutions Diversey Care Pan-Clean™ Detergent, Manual, f/Pots/Pans, J-Fill, 2.5L, 2/CT, Blue
SDP
Catalog No. 5000790166
Encompass_Preferred
Suma Pan-Clean D1.5 quickly cuts through grease on even the toughest soils while you're handwashing pots, pans, utensils, glassware and other food department equipment. Noncorrosive, highly concentrated, pot and pan detergent delivers superior grease-cutting and leaves equipment and utensils thoroughly clean. Long-lasting suds provide outstanding cleaning performance. Specially formulated detergent provides excellent cleaning in a three-compartment sink and leaves sink areas smelling clean and fresh with a floral scent. Dilution ratio is 1:600. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.
Catalog No. 50-007-90166 Supplier Enterprise Technology Solutions Supplier No. DVO100839776
Suma Pan-Clean D1.5 quickly cuts through grease on even the toughest soils while you're handwashing pots, pans, utensils, glassware and other food department equipment. Noncorrosive, highly concentrated, pot and pan detergent delivers superior grease-cutting and leaves equipment and utensils thoroughly clean. Long-lasting suds provide outstanding cleaning performance. Specially formulated detergent provides excellent cleaning in a three-compartment sink and leaves sink areas smelling clean and fresh with a floral scent. Dilution ratio is 1:600. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.