Learn More
Enterprise Technology Solutions Diversey Care Rinse Aid,f/Dishwashers,Low-Foam,Quick-Dry,Spot-Free,Streak-Free,1 Gallon,4/CT,Blue
SDP
Catalog No. 5000830920
Encompass_Preferred
Suma Rinse A5 is an economy rinse aid for soft-to-medium water conditions. Special low-foam formula can be used in a wide range of warewashing machines to ensure quick drying. A special blend of nonionic surfactants deliver rapid streak-free and spot-free results.
Catalog No. 50-008-30920 Supplier Enterprise Technology Solutions Supplier No. DVO57268280
Suma Rinse A5 is an economy rinse aid for soft-to-medium water conditions. Special low-foam formula can be used in a wide range of warewashing machines to ensure quick drying. A special blend of nonionic surfactants deliver rapid streak-free and spot-free results.