Learn More
Enterprise Technology Solutions Diversey Care Sanitizer,Optifill,Concentrate,No Rinse,2.5L,Red
SDP
Catalog No. 5000789881
Encompass_Preferred
No-rinse sanitizer conveniently and effectively sanitizes food-contact surfaces where sanitization is of prime importance. Use Final Step Sanitizer in public eating places and commercial sinks for sink immersion of utensils as well as on dairy processing and food-processing equipment. It yields a concentration of 200 ppm active quat. With the dilution control system of Optifill, the job of sanitizing is done thoroughly and correctly every time. Sanitizer with a quaternary scent is Kosher certified and NSF D2 listed.
Catalog No. 50-007-89881 Supplier Enterprise Technology Solutions Supplier No. DVO101105267
No-rinse sanitizer conveniently and effectively sanitizes food-contact surfaces where sanitization is of prime importance. Use Final Step Sanitizer in public eating places and commercial sinks for sink immersion of utensils as well as on dairy processing and food-processing equipment. It yields a concentration of 200 ppm active quat. With the dilution control system of Optifill, the job of sanitizing is done thoroughly and correctly every time. Sanitizer with a quaternary scent is Kosher certified and NSF D2 listed.