Learn More
Enterprise Technology Solutions Diversey Care Stainless Steel Cleaner/Polish,Aerosol, 482g,12/CT,Lemon
SDP
Catalog No. 5000831312
Encompass_Preferred
Stainless steel cleaner and polish removes grease, dirt, fingerprints, smudges and water marks from stainless steel, brass, aluminum and chrome. Water-based, aerosol formula leaves no residue or film. Twinkle Cleaner and Polish has a light lemon scent that's pleasant to use yet it is odorless after polishing. It contains no CFCs and is NSF C1 Registered.
Catalog No. 50-008-31312 Supplier Enterprise Technology Solutions Supplier No. DVO991224CT
Stainless steel cleaner and polish removes grease, dirt, fingerprints, smudges and water marks from stainless steel, brass, aluminum and chrome. Water-based, aerosol formula leaves no residue or film. Twinkle Cleaner and Polish has a light lemon scent that's pleasant to use yet it is odorless after polishing. It contains no CFCs and is NSF C1 Registered.