Learn More
Enterprise Technology Solutions Diversey Care Suma Grill/Oven Cleaner, Gel, 32 oz, 12/CT, Tan
SDP
Catalog No. 5000790733
Encompass_Preferred
Suma Oven and Grill Cleaner D9.6 is a quality, thickened ready-to-use grill and oven cleaner. Highly alkaline, fast-acting detergent cleans heavily soiled ovens and grills. It can also be used on porcelain. Cleaner easily cuts through baked-on grease and carbon in ovens, grills and deep fryers. This blend of alkali and surfactants removes a wide range of soils and clings to vertical surfaces. Thick gel maximizes contact time.
Catalog No. 50-007-90733 Supplier Enterprise Technology Solutions Supplier No. DVO948049
Suma Oven and Grill Cleaner D9.6 is a quality, thickened ready-to-use grill and oven cleaner. Highly alkaline, fast-acting detergent cleans heavily soiled ovens and grills. It can also be used on porcelain. Cleaner easily cuts through baked-on grease and carbon in ovens, grills and deep fryers. This blend of alkali and surfactants removes a wide range of soils and clings to vertical surfaces. Thick gel maximizes contact time.