Learn More
Enterprise Technology Solutions Diversey Care Suma INOX D7, Stainless Steel Polish, 946ml, 6/CT, CL
SDP
Catalog No. 5000831314
Encompass_Preferred
Suma INOX D7 is an oil-based, ready-to-use cleaner and polish for use on stainless steel, nonfood contact surfaces and other metal surfaces, such as brass, aluminum and copper. Ideal for use on refrigerator doors, handles, trolleys and dishwashing machines, this cleaner leaves the surface clean and shiny. It helps protect surfaces by forming a protective film, keeping surfaces looking good with a smooth luster. It resists finger marks and water spotting to maintain a polished appearance for longer. Cleaner is colorless and has a hydrocarbon scent.
Catalog No. 50-008-31314 Supplier Enterprise Technology Solutions Supplier No. DVO94368259CT
Suma INOX D7 is an oil-based, ready-to-use cleaner and polish for use on stainless steel, nonfood contact surfaces and other metal surfaces, such as brass, aluminum and copper. Ideal for use on refrigerator doors, handles, trolleys and dishwashing machines, this cleaner leaves the surface clean and shiny. It helps protect surfaces by forming a protective film, keeping surfaces looking good with a smooth luster. It resists finger marks and water spotting to maintain a polished appearance for longer. Cleaner is colorless and has a hydrocarbon scent.