Learn More
Enterprise Technology Solutions Diversey Care UHS SC Floor Cleaner/Maintainer, Concentrated, Accumix 4, 32oz, 6/CT, Green
SDP
Catalog No. 5000790903
Encompass_Preferred
UHS SC Floor Cleaner/Maintainer delivers high-performance, low-foam cleaning without rinsing and leaves no dulling residue. It keeps floor finishes cleaner longer, extending stripping cycles and saving time and labor. Super-concentrated cleaner is specifically formulated to clean burnished, ultra high-speed floor finishes. Ideal for automatic scrubbers, low-foam formula requires less emptying of recovery tanks for more machine time on the floor. Dilution ratio range is 1:256 to 1:1024. For an accurate solution to cleaner dilution, AccuMix Portion Control System is a unique cleaning system with a built-in, accurate measuring spout. Simply loosen the cap, squeeze and pour the concentrate into a bucket or scrubber tank. You get the right proportions every time. No measuring hassles mean less product waste.
Catalog No. 50-007-90903 Supplier Enterprise Technology Solutions Supplier No. DVO94529489
UHS SC Floor Cleaner/Maintainer delivers high-performance, low-foam cleaning without rinsing and leaves no dulling residue. It keeps floor finishes cleaner longer, extending stripping cycles and saving time and labor. Super-concentrated cleaner is specifically formulated to clean burnished, ultra high-speed floor finishes. Ideal for automatic scrubbers, low-foam formula requires less emptying of recovery tanks for more machine time on the floor. Dilution ratio range is 1:256 to 1:1024. For an accurate solution to cleaner dilution, AccuMix Portion Control System is a unique cleaning system with a built-in, accurate measuring spout. Simply loosen the cap, squeeze and pour the concentrate into a bucket or scrubber tank. You get the right proportions every time. No measuring hassles mean less product waste.