Learn More
Enterprise Technology Solutions Diversey Care Urinal Screen, Citrus, 7-3/4"Wx8"Lx7-1/4"H, 12/CT, Yellow
SDP
Catalog No. 5000790579
Encompass_Preferred
Urinal screen features an anti-splash design that uses sealed bristle technology to help prevent splashback to your walls and floors. Flexible plastic conforms to any size or shape urinal to provide a perfect fit. Urinal screen is infused with a proprietary resin that uses time-release deodorant technology to keep the area smelling fresh for at least 60 days. Urinal screen is designed for high-traffic public restrooms, such as those found in hotels, convention centers, restaurants and hospitals.
Catalog No. 50-007-90579 Supplier Enterprise Technology Solutions Supplier No. DVOEKS13C12
Urinal screen features an anti-splash design that uses sealed bristle technology to help prevent splashback to your walls and floors. Flexible plastic conforms to any size or shape urinal to provide a perfect fit. Urinal screen is infused with a proprietary resin that uses time-release deodorant technology to keep the area smelling fresh for at least 60 days. Urinal screen is designed for high-traffic public restrooms, such as those found in hotels, convention centers, restaurants and hospitals.