Learn More
Enterprise Technology Solutions Diversey Care WipeSystem,w/Bucket,Disposab,12"x10.5",125Wipes/Tub,6/CT,WE
SDP
Catalog No. 5000790285
Encompass_Preferred
EasyWipe Disposable Wiping System provides a pre-wetted wiping system for flexibility and ease. Simply add your cleaner, disinfectant or sanitizer of choice to pre-moisten disposable wipes. Specially treated wipes do not bind with cleaning chemicals or disinfectant actives and can be used with a variety of Diversey products. High-quality fibers allow easy pick-up and removal of dirt, dust and other soils. Individual wipes are in an enclosed system to reduce the risk of surface contamination, chemical splashes and spills. Buckets can be re-used with an EasyWipe refill (not included). Use wipes on all hard, nonporous surfaces that are outlined on the chemical label.
Catalog No. 50-007-90285 Supplier Enterprise Technology Solutions Supplier No. DVO5768748
EasyWipe Disposable Wiping System provides a pre-wetted wiping system for flexibility and ease. Simply add your cleaner, disinfectant or sanitizer of choice to pre-moisten disposable wipes. Specially treated wipes do not bind with cleaning chemicals or disinfectant actives and can be used with a variety of Diversey products. High-quality fibers allow easy pick-up and removal of dirt, dust and other soils. Individual wipes are in an enclosed system to reduce the risk of surface contamination, chemical splashes and spills. Buckets can be re-used with an EasyWipe refill (not included). Use wipes on all hard, nonporous surfaces that are outlined on the chemical label.