Learn More
MSC Diversey Extraction Rinse, Liquid, 1 Gal, Floral Scent, pH 3.19
Catalog No. 502839147
Encompass
Diversey Extraction Rinse is a liquid carpet cleaner with a floral scent, formulated for use in carpet and upholstery extraction cleaning. This product helps to remove hard water deposits and neutralize alkaline residues from prior cleaning treatments, leaving carpets clean and fresh. It is intended to be diluted before use.
  • Liquid form
  • 1 gallon container size
  • Floral scent
  • Designed for carpet and upholstery cleaning
  • Undiluted pH of approximately 3.19
  • Completely soluble
  • Clear colorless liquid
  • Contains an added fragrance
Catalog No. 50-283-9147 Supplier MSC Supplier No. 50544196
Diversey Extraction Rinse is a liquid carpet cleaner with a floral scent, formulated for use in carpet and upholstery extraction cleaning. This product helps to remove hard water deposits and neutralize alkaline residues from prior cleaning treatments, leaving carpets clean and fresh. It is intended to be diluted before use.
  • Liquid form
  • 1 gallon container size
  • Floral scent
  • Designed for carpet and upholstery cleaning
  • Undiluted pH of approximately 3.19
  • Completely soluble
  • Clear colorless liquid
  • Contains an added fragrance