Learn More
MSC Diversey Spray Disinfectant, Aerosol, 15 oz, 12/Carton
Catalog No. 502868398
Encompass
This quaternary-based aerosol germicide is designed for disinfecting pre-cleaned surfaces. It is fungicidal and tuberculocidal, effectively controlling mold and mildew. Suitable for use in various environments like hospital rooms, sick rooms, and locker room facilities on hard, non-porous environmental surfaces.
  • Kills gram positive and gram negative bacteria
  • Ideal for locker rooms, shower stalls, floors, and benches
  • Kills Staphylococcus aureus, Pseudomonas aeruginosa, and Streptococcus hemolyticus on hard, non-porous environmental surfaces
  • Eliminates odors at their source
  • Controls the growth of fungus, mold, and mildew on hard, non-porous environmental surfaces
Catalog No. 50-286-8398 Supplier MSC Supplier No. 39695853
This quaternary-based aerosol germicide is designed for disinfecting pre-cleaned surfaces. It is fungicidal and tuberculocidal, effectively controlling mold and mildew. Suitable for use in various environments like hospital rooms, sick rooms, and locker room facilities on hard, non-porous environmental surfaces.
  • Kills gram positive and gram negative bacteria
  • Ideal for locker rooms, shower stalls, floors, and benches
  • Kills Staphylococcus aureus, Pseudomonas aeruginosa, and Streptococcus hemolyticus on hard, non-porous environmental surfaces
  • Eliminates odors at their source
  • Controls the growth of fungus, mold, and mildew on hard, non-porous environmental surfaces