Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Diversey Hand Soap, Foam, Fragrance Free, Antibacterial, 1.2 L, 6/Pk.

Catalog No. 502816731
Encompass

Diversey Soft Care Defend Foam Handwash is an antibacterial, fragrance-free foam hand soap designed for commercial and industrial use, suitable for high-speed applications.

  • Foam formula for effective cleaning
  • Fragrance free for sensitive users
  • Antibacterial properties for enhanced hygiene
  • Cartridge capacity of 1.2 L for convenient dispensing
  • Clear color
  • Flammable liquid and vapor
  • Has a pH of 8.5
  • Flash point 130 °F (54.5 °C)
  • Density 0.9999 Kg/L

Catalog No. 50-281-6731 Supplier MSC Supplier No. 50019744
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diversey Soft Care Defend Foam Handwash is an antibacterial, fragrance-free foam hand soap designed for commercial and industrial use, suitable for high-speed applications.

  • Foam formula for effective cleaning
  • Fragrance free for sensitive users
  • Antibacterial properties for enhanced hygiene
  • Cartridge capacity of 1.2 L for convenient dispensing
  • Clear color
  • Flammable liquid and vapor
  • Has a pH of 8.5
  • Flash point 130 °F (54.5 °C)
  • Density 0.9999 Kg/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.