Learn More
MSC Diversey Spotter Gel, P.O.G., Gel, 32 oz, RTU
Catalog No. 502835625
Encompass
Diversey P.O.G. Spotter Gel is a ready-to-use solvent-based gel designed for effective removal of paint, oil, and grease stains from carpets and upholstery. It is formulated for industrial and institutional carpet care applications.
  • Effectively removes paint, oil, and grease stains.
  • Ready to use formula.
  • Designed for industrial and institutional carpet care.
  • Gel consistency for targeted application.
  • Not corrosive to metals.
  • Available in a 32 ounce squeeze bottle.
Catalog No. 50-283-5625 Supplier MSC Supplier No. 50312669
Diversey P.O.G. Spotter Gel is a ready-to-use solvent-based gel designed for effective removal of paint, oil, and grease stains from carpets and upholstery. It is formulated for industrial and institutional carpet care applications.
  • Effectively removes paint, oil, and grease stains.
  • Ready to use formula.
  • Designed for industrial and institutional carpet care.
  • Gel consistency for targeted application.
  • Not corrosive to metals.
  • Available in a 32 ounce squeeze bottle.