Learn More
Waters Corp Diverter Rotor/Stator Kit, Rebuild Kit, MS System
Catalog No. 502391276 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides a replacement rotor and stator for a diverter valve pod. It is an essential component of the fluidics assembly for various Mass Spectrometers, including SQD2, XevoTQD, Xevo TQ-S micro, and Xevo TQ-S cronos systems.
  • Contains a replacement rotor and stator.
  • Functions as a rebuild kit.
  • Designed for MS system types.
  • Replaced as required for maintenance.
Catalog No. 50-239-1276 Supplier Waters Corp Supplier No. 700009465
This kit provides a replacement rotor and stator for a diverter valve pod. It is an essential component of the fluidics assembly for various Mass Spectrometers, including SQD2, XevoTQD, Xevo TQ-S micro, and Xevo TQ-S cronos systems.
  • Contains a replacement rotor and stator.
  • Functions as a rebuild kit.
  • Designed for MS system types.
  • Replaced as required for maintenance.