Learn More
Waters Corp Waters Solenoid Diverter Valve, 100 psi, 3-Way, 2-Position
Catalog No. 502039219 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit includes a solenoid diverter valve designed for fluidic control in analytical instrumentation. It provides precise control for fluidic pathways and is rated for 100 psi.
  • Solenoid diverter valve
  • 2-position, 3-way configuration
  • Rated for 100 psi
  • Includes ETFE tubing for flow rate selections
  • Includes cable ties for securing tubing
  • Designed for connection to a second fraction collector
Catalog No. 50-203-9219 Supplier Waters Corp Supplier No. 205000155
This kit includes a solenoid diverter valve designed for fluidic control in analytical instrumentation. It provides precise control for fluidic pathways and is rated for 100 psi.
  • Solenoid diverter valve
  • 2-position, 3-way configuration
  • Rated for 100 psi
  • Includes ETFE tubing for flow rate selections
  • Includes cable ties for securing tubing
  • Designed for connection to a second fraction collector