Learn More
Waters Corp Diverter Valve Pod, Mass Spectrometry, Spare Parts
Catalog No. 502392238 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement Diverter Valve Pod is an essential component for maintaining the optimal performance of Mass Spectrometer systems.
  • Specifically designed for use with the diverter valve found on SQD2, Xevo TQD, Xevo TQ-S micro, and Xevo TQ-S cronos mass spectrometers.
  • Classified as a spare parts kit, ensuring necessary components for critical replacements.
  • Valve component.
  • Designed for mass spectrometry (MS) systems.
  • Recommended for replacement as required, allowing for responsive maintenance to minimize downtime.
Catalog No. 50-239-2238 Supplier Waters Corp Supplier No. 700009462
This replacement Diverter Valve Pod is an essential component for maintaining the optimal performance of Mass Spectrometer systems.
  • Specifically designed for use with the diverter valve found on SQD2, Xevo TQD, Xevo TQ-S micro, and Xevo TQ-S cronos mass spectrometers.
  • Classified as a spare parts kit, ensuring necessary components for critical replacements.
  • Valve component.
  • Designed for mass spectrometry (MS) systems.
  • Recommended for replacement as required, allowing for responsive maintenance to minimize downtime.