Learn More
Uline Divider Box - 15 x 9 x 3"", Blue
Catalog No. NC2543397
Encompass
This item must be ordered in multiples of 12 per supplier requirements
Customize storage space with these dividable boxes.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.
Catalog No. NC2543397 Supplier Uline Supplier No. S19495BLUPK
Add to Cart
Add to Cart
This item must be ordered in multiples of 12 per supplier requirements
Customize storage space with these dividable boxes.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.