Learn More
Uline Divider Box - 20 x 15 x 6", Clear, Customize storage space with these dividable boxes.*****Must be ordered in multiples of 3*****
Catalog No. NC2880726
Encompass
Customize storage space with these dividable boxes.Spot what you need quickly.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.Optional dividers sold separately.
Catalog No. NC2880726 Supplier Uline Supplier No. S23362
Customize storage space with these dividable boxes.Spot what you need quickly.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.Optional dividers sold separately.