Learn More
Uline DIVIDER BOX GR 20X15X2.5 6/PK
Catalog No. NC1755788
Encompass
Gray, 6 pk.Use in stock rooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.
Catalog No. NC1755788 Supplier Uline Supplier No. S22734GRCS
Gray, 6 pk.Use in stock rooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.