Learn More
Uline Divider Box - 20 x 15 x 2 1⁄2", Gray
Catalog No. NC3243652
Encompass
Customize storage space with these dividable boxes.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.Optional dividers sold separately.
Catalog No. NC3243652 Supplier Uline Supplier No. S22734GR
Customize storage space with these dividable boxes.Use in stockrooms, assembly lines and repair shops.High-density polypropylene resists solvents and chemicals.Stackable. Flat panels to label contents.Optional dividers sold separately.