Learn More
Fritsch Milling And Sizing Inc Dividing Head 3 Samples, Cpl. Aluminium Ptfe-Coated, L-27
SDP
Catalog No. 501534083
Encompass
Dividing head 3 samples for LABORETTE 27 made of PTFE-coated aluminium, incl. 3 sample bottles 15 ml with screw lid and funnel 10 mm dia. as well as 1 funnel each of 5 mm dia. for dry- resp. wet division
Catalog No. 50-153-4083 Supplier Fritsch Milling And Sizing Inc Supplier No. 27.2150.00
Add to Cart
Add to Cart
Request a Quote
Dividing head 3 samples for LABORETTE 27 made of PTFE-coated aluminium, incl. 3 sample bottles 15 ml with screw lid and funnel 10 mm dia. as well as 1 funnel each of 5 mm dia. for dry- resp. wet division