Learn More
Electron Microscopy Sciences Divinyl Benzene, Mixed Isomers 100 ML
SDP
Catalog No. 50980368
Encompass_Preferred
CAS #1321-74-0 C₆H₄(CH:CH₂)₂ F.W. 130.19
A cross-linking monomer.Inhibited with up to 1,500 ppm p-tert-butyl catechol. Specific Gravity: 0.91
Catalog No. 50-980-368 Supplier Electron Microscopy Sciences Supplier No. 13450
CAS #1321-74-0 C₆H₄(CH:CH₂)₂ F.W. 130.19
A cross-linking monomer.Inhibited with up to 1,500 ppm p-tert-butyl catechol. Specific Gravity: 0.91