Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Divinyl sulfone

Catalog No. 503126045 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6045 25 g
50-312-5719 100 g
50-312-5870 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-6045 Supplier Sigma Aldrich Supplier No. V370025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Divinyl sulfone

Specifications

CAS 77-77-0
Melting Point -26°C
Density 1.177 g/mL (at 25°C)
Boiling Point 234°C
Molecular Formula C4H6O2S
Refractive Index n20/D 1.476 (literature)
Linear Formula (CH2=CH)2 SO2
Quantity 25 g
MDL Number MFCD00008623
Synonym Vinyl sulfone
Molecular Weight (g/mol) 118.15
Percent Purity ≥96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.