Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinyl Sulfone (stabilized with HQ) 96.0+%, TCI America™
SDP

Catalog No. D0959250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0959250G 250 g
D09595G 5 g
D095925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0959250G Supplier TCI America Supplier No. D0959250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77-77-0
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00008623
InChI Key AFOSIXZFDONLBT-UHFFFAOYSA-N
Synonym divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
PubChem CID 6496
ChEBI CHEBI:53729
IUPAC Name 1-ethenylsulfonylethene
SMILES C=CS(=O)(=O)C=C

Specifications

Melting Point -26°C
Boiling Point 110°C
Quantity 250 g
UN Number 2927
Formula Weight 118.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Divinyl Sulfone (stabilized with HQ)
RTECSNumber KM7175000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.