Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Divinylbenzene

Catalog No. 1690991L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-909-91L 1 L
16-909-9250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-909-91L Supplier Sigma Aldrich Supplier No. 1690991L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Divinylbenzene (DVB) has a benzene ring in combination with meta and para isomers of DVB and ethylvinylbenzene. It acts as a monomeric unit which facilitates the preparation of polydivinylbenzene by anionic polymerization.DVB can be used as a crosslinking agent in the synthesis of silicon/carbon composite for use as an anode material for lithium ion batteries. It can also be used as a monomer to synthesize dimer particles by seed polymerization method for potential uses in electrical clutches and shock absorbers.contains 4-ethylvinylbenzene and 3-ethylvinylbenzene

Specifications

CAS 1321-74-0
Density 0.912 g/mL (at 25°C)
Boiling Point 195°C
Molecular Formula C10H10
Refractive Index n20/D 1.561 (literature)
Linear Formula C6H4(CH=CH2)2
Quantity 1 L
MDL Number MFCD01778656
Synonym DVB HP; Diethenylbenzene; Vinylstyrene
Molecular Weight (g/mol) 130.19
Percent Purity 55%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.