Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Divinyltin dichloride, Thermo Scientific™

Catalog No. AA3961318
Change view
Click to view available options
Quantity:
2 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3961318 50 g
AA3961304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3961318 Supplier Thermo Scientific Chemicals Supplier No. 3961318

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7532-85-6
Molecular Formula C4H8Cl2Sn
Molecular Weight (g/mol) 245.72
MDL Number MFCD00058728
InChI Key OFJXSWMLVIFLKC-UHFFFAOYSA-N
Synonym divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
PubChem CID 82033
IUPAC Name dichloro-bis(ethenyl)stannane
SMILES Cl.Cl.C=C[SnH2]C=C

Specifications

Boiling Point 54°C to 56°C (3 mmHg)
Quantity 50 g
UN Number UN2788
Sensitivity Moisture Sensitive
Formula Weight 243.69
Physical Form Liquid
Chemical Name or Material Divinyltin dichloride
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.