Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dixylyl disulphide | 13616-83-6 | MFCD00792370 | 274.44 g/mol | C16H18S2 | 5 G
SDP

Catalog No. 5000281661
Encompass_Preferred

Dixylyl disulphide is an organosulfur reagent (bis(2,4-dimethylphenyl) disulfide) used in biochemical and analytical research as a reference standard and as a synthetic reagent. Supplied for laboratory research use only, the product should be handled according to its safety data sheet and appropriate laboratory protocols.

  • Organosulfur reagent for research and analytical applications.
  • Useful as an impurity reference standard in analytical methods.
  • Molecular formula C16H18S2 and molecular weight 274.44 g/mol.
  • CAS number 13616-83-6 for unambiguous identification.
  • Available in 5 G pack size for laboratory use.
  • Consult the safety data sheet for hazards, handling, and storage.

Catalog No. 50-002-81661 Supplier Medchemexpress LLC Supplier No. HYW3949355G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dixylyl disulphide is an organosulfur reagent (bis(2,4-dimethylphenyl) disulfide) used in biochemical and analytical research as a reference standard and as a synthetic reagent. Supplied for laboratory research use only, the product should be handled according to its safety data sheet and appropriate laboratory protocols.

  • Organosulfur reagent for research and analytical applications.
  • Useful as an impurity reference standard in analytical methods.
  • Molecular formula C16H18S2 and molecular weight 274.44 g/mol.
  • CAS number 13616-83-6 for unambiguous identification.
  • Available in 5 G pack size for laboratory use.
  • Consult the safety data sheet for hazards, handling, and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.