Learn More
Medchemexpress LLC HY-15084A 10mg Medchemexpress, (-)-Dizocilpine (Maleate) CAS:121917-57-5 Purity:>98%
SDP
Catalog No. 501932723
Encompass_Preferred
Medchemexpress, HY-15084A 10mg (-)-Dizocilpine (Maleate) CAS:121917-57-5 (-)-Dizocilpine ((-)-MK 801) Maleate is the enantiomer of (+)-MK-801. (+)-MK 801 Maleate is a potent, selective and non-competitive NMDA receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2723 Supplier Medchemexpress LLC Supplier No. HY15084A10MG
Medchemexpress, HY-15084A 10mg (-)-Dizocilpine (Maleate) CAS:121917-57-5 (-)-Dizocilpine ((-)-MK 801) Maleate is the enantiomer of (+)-MK-801. (+)-MK 801 Maleate is a potent, selective and non-competitive NMDA receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.