Learn More
TARGETMOL CHEMICALS INC --DIZOCILPINE MALEATE 50MG
SDP
Catalog No. 502576526
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Dizocilpine maleate ((-)-MK 801 (Maleate)) is a potent selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes. purity: 99%
Catalog No. 50-257-6526 Supplier TARGETMOL CHEMICALS INC Supplier No. T635250MG
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (-)-Dizocilpine maleate ((-)-MK 801 (Maleate)) is a potent selective and non-competitive NMDA receptor antagonist with Kd of 37.2 nM in rat brain membranes. purity: 99%