Learn More
TARGETMOL CHEMICALS INC DIZOCILPINE MALEATE 50MG
SDP
Catalog No. 502580318
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dizocilpine Maleate (MK 801) is a potent noncompetitive antagonist of the NMDA receptor (RECEPTORS N-METHYL-D-ASPARTATE) with Kd of 37.2 nM in rat brain membranes. The drug has been considered for the wide variety of neurodegenerative conditions or disorders in which NMDA receptors may play an important role. purity: 99%
Catalog No. 50-258-0318 Supplier TARGETMOL CHEMICALS INC Supplier No. T332050MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dizocilpine Maleate (MK 801) is a potent noncompetitive antagonist of the NMDA receptor (RECEPTORS N-METHYL-D-ASPARTATE) with Kd of 37.2 nM in rat brain membranes. The drug has been considered for the wide variety of neurodegenerative conditions or disorders in which NMDA receptors may play an important role. purity: 99%