Learn More
Selleck Chemical LLC (-)-Dizocilpine (MK 801) Maleate S2857-50mg
SDP
Catalog No. 503111862
Encompass_Preferred
(-)-Dizocilpine (MK 801 Dizocilpine C13737) Maleate is a potent N-methyl-D-aspartate (NMDA) receptor antagonist with Ki of 30 5 nM
Catalog No. 50-311-1862 Supplier Selleck Chemical LLC Supplier No. S285750MG
(-)-Dizocilpine (MK 801 Dizocilpine C13737) Maleate is a potent N-methyl-D-aspartate (NMDA) receptor antagonist with Ki of 30 5 nM